missing translation for 'onlineSavingsMsg'
Learn More

Hopen™ Iso Air Cleanroom Masks with Ties
GREENER_CHOICE

ISO Air cleanroom masks have long ties for easy donning and are made with the Mass Balance approach from recycled materials.

115.00 €

Specifications

Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Style Pleated
Certifications/Compliance Grade B
View More Specs

Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17734789
GREENER_CHOICE
View Documents
Hopen™
60.720M
Medium
115.00 €
Pack of 300
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
 
17724789
GREENER_CHOICE
View Documents
Hopen™
60.720L
Large
115.00 €
Pack of 300
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
Description

Description

  • Low particle release: Helmke drum tested for use in clean rooms
  • High filtration efficiency of particles, bacteria and viruses
  • Adaptable to users and protocols with two sizes available
  • Headloops improve comfort for the user by making it easier to put on
  • Softex technology limits risk of particulate contamination while reducing risk of skin irritation for the user
  • Adjustable and malleable nose clip prevents glasses from fogging up
  • ISO AIR range is manufactured in an ISO 8 classified clean room
  • Made in France
Specifications

Specifications

Cleanroom Face Mask
Ties
Grade B
>99%
60.720
White
Pleated
Class 100 (ISO 5)
Industry
Videos
SDS
Documents

Documents

Product Certifications