missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hopen™ Iso Air Duckbill Cleanroom Masks
ISO Air duckbill cleanroom masks provide a large breathing chamber and are made with the Mass Balance approach from recycled materials.
Brand: Hopen™ 60.754L
This item is not returnable.
View return policy
Description
- Low particle release: Helmke drum tested for use in clean rooms
- High filtration efficiency of particles, bacteria and viruses
- Adaptable to users and protocols with two sizes available
- Highly breathable thanks to a large breathing chamber
- Softex technology limits risk of particulate contamination while reducing risk of skin irritation for the user
- Adjustable and malleable nose clip prevents glasses from fogging up
- ISO AIR range is manufactured in an ISO 8 classified clean room
- Made in France
Specifications
Cleanroom Face Mask | |
Large | |
Grade B | |
Industry |
White | |
Pouch | |
Class 1 (ISO 3) | |
60.754 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction