missing translation for 'onlineSavingsMsg'
Learn More

iso-Hexane, BAKER ANALYZED HPLC, J.T. Baker™

Product Code. 10548531
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Get rapid, reproducible performance and separation results. iso-Hexane, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use

Specifications

Melting Point 0°C
Absorbance Max. 0.2 at 210nm, Max. 0.08 at 220nm, Max. 0.008 at 254nm, Max. 0.005 at 280nm, Max. 0.005 at 354nm
CAS 64742-49-0
Color Colorless
Density 0.7g/cm3
Boiling Point 57°C to 63°C
Quantity 2.5 L
Flash Point −33°C
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 0.0005%
Molecular Formula C6H14
MDL Number MFCD00081849
Water Max. 0.01%
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
Solubility Information Not or partly miscible in water
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES CCCC(C)C
IUPAC Name 2-methylpentane
Molecular Weight (g/mol) 86.178
PubChem CID 7892
Formula Weight 86.18g/mol
Percent Purity ≥95%
Grade For use in High Performance Liquid Chromatography
Chemical Name or Material iso-Hexane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.