missing translation for 'onlineSavingsMsg'
Learn More

iso-Octane, Honeywell Burdick & Jackson™

Product Code. 16655903
Click to view available options
200 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

Melting Point -107°C
CAS 540-84-1
Color Colorless
CAS Max % 1
Density 0.69 g/cm3 (20°C)
Boiling Point 99°C (1013 hPa)
Quantity 200 L
Flash Point -12°C
Molecular Formula C8H18
Linear Formula CH3(CH2)6CH3
Beilstein 1696876
Synonym Isooctane, 2,2,4-Trimethylpentane
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Molecular Weight (g/mol) 114.23
Grade ACS Plus
Chemical Name or Material iso-Octane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'