missing translation for 'onlineSavingsMsg'
Learn More

iso-Propyl Ether, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15548554
Click to view available options
Quantity:
1 L
Packaging:
Aluminum Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Flash Point −28.00°C
Packaging Aluminum Bottle
Quantity 1 L
Specific Gravity 1L = 0.78kg
Formula Weight 102.18
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material iso-Propyl Ether
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.