missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ISO Safety Sign without Text - Warning: Biological Hazard
Safety sign for hazard identification.
Brand: Brady™ 139020
10.58 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
Gloss | |
Permanent Acrylic | |
9 x 10 mm |
Laminated Polyester | |
Triangular | |
Black on Yellow |