missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ISO Safety Sign without Text - Warning: Magnetic Field
Safety sign for hazard identification.
Brand: Brady™ 827203
9.24 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
PIC W006-Tri 025-PE-SHEET/13 | |
Laminated Polyester | |
Triangular | |
Black on Yellow |
Gloss | |
Permanent Acrylic | |
22 x 25 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction