Learn More
Brady™ ISO Square Polyester Safety Sign 'Fire Alarm Flashing Light'
ISO Safety Sign displaying flashing fire alarm and flames.
Brand:  Brady™ 311199
 View more versions of this product
Product Code. 17841795
  • 3.45 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Specifications
F/F018/NT/PE-100X100-1
Polyester
Permanent
Facility Identification, Safety Identification
3.94 in.
Square
Universal
Fire Alarm Flashing Light
White on Red
Gloss
1 Sign
Surface Mount
3.94 in.
-40°C to +110°C
-40°F to +230°F
100 x 100 mm
Safety Sign