missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alla France™ ISO Standard Hydrometer
Use these hydrometers for a variety of applications. Alla France™ ISO Standard Hydrometer feature a benchmark engraved on the stem to show any displacement of the scale.
49.65 €
Product Code | Brand | Density (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Density (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
11753423
|
Alla France™
3100MC130/20/H-qp |
0.130 to 0.140g/mL | 250 mm |
49.65 €
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
Description
- Density Hydrometers (g/mL)
- ISO, BS, DIN, NF... MRA Traceable
- General Purpose at 20°C
- The scale of the hydrometer is fully marked with all the reference norms, the calibration temperature, the surface tension, the individual number of the instrument, the serial number and the ALLA FRANCE logo
- Reading below meniscus.
- Systematically delivered with a Traceability and Conformity Certificate.
- On request, can be supplied with “EU” Traceable Calibration Certificate or “EO” Official Calibration Certificate
- from 0.600 to 1.000 g/mL= 16 at 35 mN/m (low tension)
- from 1.000 to 1.300 g/mL= 55 mN/m (medium tension)
- from 1.300 to 2.000 g/mL= 75 mN/m (high tension)
Specifications
1Each | |
ISO Standard Hydrometer |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Alla France™ ISO Standard Hydrometer