missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Alcohol, MP Biomedicals™
Click to view available options
Quantity:
500 mL
Specifications
Specifications
CAS | 123-51-3 |
Melting Point | -117.2°C (literature) |
Density | 0.809g/mL at 20°C(Lit.) |
Boiling Point | 131°C (literature) |
Molecular Formula | C5H12O |
Quantity | 500 mL |
Synonym | 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol |
InChI Key | PHTQWCKDNZKARW-UHFFFAOYSA-N |
SMILES | CC(C)CCO |
IUPAC Name | 3-methylbutan-1-ol |
Show More |
Safety and Handling
Product Identifier
- ISOAMYL ALCOHOL 500ML
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : isoamyl alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction