missing translation for 'onlineSavingsMsg'
Learn More
Isobutanol, 99%, Thermo Scientific Chemicals Product Code.: 11466286

Isobutanol, 99%, Thermo Scientific Chemicals

Product Code. 11466286
500 mL
Click to view available options
Quantity:
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11466286

Brand: Thermo Scientific Alfa Aesar B23091.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.

Solubility
Miscible with alcohol and diethyl ether. Partially miscible with water.

Notes
Avoid all sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcoholShow MoreShow Less
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

pH 7
Melting Point -108°C
Density 0.802
Boiling Point 107°C to 108°C
Quantity 500 mL
Flash Point 28°C (82°F)
Odor Sweet
Refractive Index 1.396
Linear Formula (CH3)2CHCH2OH
UN Number UN1212
Beilstein 1730878
Merck Index 14,5131
Solubility Information Miscible with alcohol and diethyl ether. Partially miscible with water.
Formula Weight 74.12
Percent Purity 99%
Chemical Name or Material Isobutanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251095 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB23091.AP,B23091.0F
Lot Number10249499 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog NumberB23091.AP,B23091.0F
Lot Number10248397 Certificate TypeCertificate of Analysis Date23/10/2023 Catalog NumberB23091.AP,B23091.0F
Lot NumberW28I021 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberB23091.AP,B23091.0F
Lot Number10241366 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberB23091.AP,B23091.0F
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isobutanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Isobutyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isobutanol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.