missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine, 99%, Thermo Scientific Chemicals

26.33 € - 73.47 €

Chemical Identifiers

CAS 78-81-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00008146
InChI Key KDSNLYIMUZNERS-UHFFFAOYSA-N
Synonym isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
PubChem CID 6558
ChEBI CHEBI:15997
IUPAC Name 2-methylpropan-1-amine
SMILES CC(C)CN
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498843
View Documents
Thermo Scientific Alfa Aesar
A11017.AK
250 mL
26.33 €
250mL
Please to purchase this item. Need a web account? Register with us today!
11488843
View Documents
Thermo Scientific Alfa Aesar
A11017.AU
1000 mL
73.47 €
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Isobutylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutylamine is used as pharmaceutical intermediate.

Solubility
Soluble in water (1000 mg/ml at 25°C), ether, alcohol, acetone and benzene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications

Chemical Identifiers

78-81-9
73.139
KDSNLYIMUZNERS-UHFFFAOYSA-N
6558
2-methylpropan-1-amine
C4H11N
MFCD00008146
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
CHEBI:15997
CC(C)CN

Specifications

78-81-9
0.736 g/mL
-9°C (15°F)
C4H11N
MFCD00008146
385626
14,5132
Soluble in water (1000mg/ml at 25°C),ether,alcohol,acetone and benzene.
CC(C)CN
73.139
CHEBI:15997
99%
Isobutylamine
°C to 85°C
67°C to 69°C
99%
1.397
UN1214
Air Sensitive
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
KDSNLYIMUZNERS-UHFFFAOYSA-N
2-methylpropan-1-amine
6558
73.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H314-H318

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H301-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYLAMINE

missing translation for 'einecsNumber' : 201-145-4

missing translation for 'rtecsNumber' : NP9900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • Isobutylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Isobutylamine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.