missing translation for 'onlineSavingsMsg'
Learn More
Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10603405

Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10603405
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10603405

Brand: Thermo Scientific Acros 427391000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-62-5, 60-29-7
Molecular Formula C4H9BrMg
Molecular Weight (g/mol) 161.33
MDL Number MFCD00075126
InChI Key CMWBEISSZHZIMU-UHFFFAOYSA-M
Synonym isobutylmagnesium bromide, bromo 2-methylpropyl magnesium, magnesium,bromo 2-methylpropyl, isobutylmagnesiumbromide, bromo isobutyl magnesium, iso-butymagnesium bromide, isobutyl magnesium bromide, isobutyl-magnesium bromide, iso-butyl magnesium bromide, 2-methylpropylmagnesiumbromide, grignard reagent
PubChem CID 4285172
IUPAC Name magnesium;2-methanidylpropane;bromide
SMILES CC(C)C[Mg]Br

Specifications

Density 0.9410g/mL
Boiling Point 34.6°C
Flash Point −34°C
Packaging AcroSeal™ Glass bottle
Linear Formula (CH3)2CHCH2MgBr
Quantity 100 mL
Specific Gravity 0.941
Formula Weight 161.32
Chemical Name or Material Isobutylmagnesium bromide, 2M solution in diethyl ether

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0412547 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number427391000,427398000
Lot NumberA0416441 Certificate TypeCertificate of Analysis Date07/02/2023 Catalog Number427391000,427398000
Lot NumberA0410573 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number427391000,427398000
Lot NumberA0383003 Certificate TypeCertificate of Analysis Date28/09/2020 Catalog Number427391000,427398000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isobutylmagnesium bromide, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.