missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Sterile Bouffant with Elastic Headband
BIOPHARMA

Product Code. 15674904
Click to view available options
Size: Universal
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. DuPont™ Tyvek™ IsoClean™ Sterile Bouffant with Elastic Headband provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and low linting.

  • Clean-processed and gamma-sterilized.
  • Elastic headband.
  • One Size fits most
  • Available in white
TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Cleanroom Bouffant
Size Universal
Material Flashspun HDPE
Color White
Disposable Single-use
Model D15466053
Sterility Sterile
Certifications/Compliance ISO 11137-1, PPE Category 1
Packaging Type Individually Packed
Quantity 250/Pk.
Construction Flashspun HDPE
For Use With (Application) Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics Industry
Mostrar más Mostrar menos

The user should make the risk assessment to determine the protection required.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'