missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ IsoClean™ Unhooded Coverall with Bound Neck Product Code.: 15694894
Estimated Shipment: 20-05-2025
to see stock.
Add to basket

DuPont™ Tyvek™ IsoClean™ Unhooded Coverall with Bound Neck
BIOPHARMA

Product Code. 15694894 Shop All DuPont Products
Small
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy

Product Code. 15694894

Brand: DuPont™ D15429648

Estimated Shipment: 20-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. DuPont™ Tyvek™ IsoClean™ Unhooded Coverall with Bound Neck provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and low linting.

  • Available in sizes S to XXXL
  • Clean-processed and gamma-sterilized
  • Bound internal seams. Elasticated waist at back for good fit
  • Tyvek™ covered elasticated thumb loops
  • Tunneled elastication at wrists and ankles
  • Front zipper closure
  • Storm flap
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Small
Material HDPE
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper
Seam Type Bound
Sterility Sterile
Certifications/Compliance ISO 11137-1, ISO Class 5
Certifications/Compliance (Europe) GMP class A/B, Category III, Type 5-B and 6-B, EN 14126, EN 1073-2
Model D15429648
Size (Metric) Chest 84 to 92 cm
For Use With (Application) Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics Industry. and Other Critical Environments
Quantity 25/Pk.
Show More Show Less
  • Dupont Tyvek - Packaging for CE certified IsoClean garments
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Dupont Tyvek Packaging for CE certified IsoClean garments

    • Dupont Tyvek - IsoClean design
      Video Player is loading.
      Current Time 0:00
      Duration 1:35
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 1:35
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected

      Dupont Tyvek IsoClean design

    • DuPont Tyvek - IsoClean
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        DuPont Tyvek IsoClean

      Brochures

      Posters

      Product Information

      Certificates

      A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

      5 results found
      Lot Number Certificate Type Date Catalog Number
      Lot NumberAHS2444138 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15429648
      Lot NumberAHS2331136 Certificate TypeCertificate of Sterility Date05/09/2023 Catalog NumberD15429648
      Lot NumberAHS2331135 Certificate TypeCertificate of Sterility Date05/09/2023 Catalog NumberD15429648
      Lot NumberAHS2327162 Certificate TypeCertificate of Sterility Date24/08/2023 Catalog NumberD15429648
      Lot NumberAHS2322144 Certificate TypeCertificate of Sterility Date24/08/2023 Catalog NumberD15429648

      The user should make the risk assessment to determine the protection required.

      Product Content Correction

      Your input is important to us. Please complete this form to provide feedback related to the content on this product.

      Product Title
      DuPont™ Tyvek™ IsoClean™ Unhooded Coverall with Bound Neck >

      By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

      Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.