missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isohexane, contains <5% N-Hexane, Extra Pure, SLR
49.05 € - 585.00 €
Chemical Identifiers
CAS | 73513-42-5 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD00009406 |
InChI Key | AFABGHUZZDYHJO-UHFFFAOYSA-N |
Synonym | isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane |
PubChem CID | 7892 |
IUPAC Name | 2-methylpentane |
SMILES | CCCC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10779234
|
Fisher Chemical
H/0401/15 |
1 L | Amber Glass Bottle |
49.05 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10468980
|
Fisher Chemical
H/0401/PB17 |
2.5 L | Plastic-coated Glass Bottle |
70.75 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10396370
|
Fisher Chemical
H/0401/21 |
5 L | Metal Can |
158.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11488343
|
Fisher Chemical
H/0401/24 |
10 L | Metal Can |
211.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10181900
|
Fisher Chemical
H/0401/25 |
25 L | Metal Can |
585.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
73513-42-5 | |
86.18 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
7892 | |
CCCC(C)C |
C6H14 | |
MFCD00009406 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
2-methylpentane |
Specifications
-154°C | |
Colorless | |
Liquid | |
MFCD00009406 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
2-methylpentane | |
7892 |
73513-42-5 | |
60°C | |
C6H14 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
CCCC(C)C | |
86.18 | |
86.18g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Isohexane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title