missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isohexane, contains <5% N-Hexane, for HPLC
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Plastic-coated Glass Bottle
Specifications
Specifications
Melting Point | -154°C |
Color | Colorless |
Boiling Point | 60°C |
Quantity | 5 L |
Packaging | Aluminum Can |
Physical Form | Liquid |
Synonym | isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane |
Formula Weight | 86.18g/mol |
Safety and Handling
Product Identifier
- Isohexane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'