missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl nitrite, 97%, stab. with 0.2% anhyd. sodium carbonate, Thermo Scientific Chemicals

Product Code. 11343307
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Isopentyl nitrite is used in the conversion of 2,6-disubstituted phenols to diphenoquinones and aminophenols to quinone diazides. It acts as an oxidant and nitrosating agent. It finds application in aromatic arylation, deamination of arylamines using tetrahydrofuran as a hydrogen donor. It is also useful for the preparation of benzyne using anthranilic acid. It is an antihypertensive and vasodilator to treat heart diseases such as angina and an antidote for cyanide poisoning. Further, it is used as a solvent and cleaning agent for a printed circuit board. In addition to this, it is considered to be a replacement for freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with methanol, chloroform and ether. Slightly miscible with water.

Notes
Air, moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and alcohol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-46-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00002057
InChI Key OWFXIOWLTKNBAP-UHFFFAOYSA-N
Synonym isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i
PubChem CID 8053
ChEBI CHEBI:2691
IUPAC Name 3-methylbutyl nitrite
SMILES CC(C)CCON=O

Specifications

Density 0.875
Boiling Point 95°C to 100°C
Flash Point −20°C (−4°F)
Odor Sweet
Refractive Index 1.387
Quantity 25 mL
UN Number UN1113
Beilstein 969510
Sensitivity Air and light sensitive
Merck Index 14,5123
Solubility Information Miscible with methanol,chloroform and ether. Slightly miscible with water.
Formula Weight 117.15
Percent Purity 97%
Chemical Name or Material Isopentyl nitrite, stabilized with 0.2% anhyd. sodium carbonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isopentyl nitrite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Isopentyl nitrite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.