missing translation for 'onlineSavingsMsg'
Learn More
Isophthalic acid, 99%, Thermo Scientific Chemicals Product Code.: 11487097

Isophthalic acid, 99%, Thermo Scientific Chemicals

Product Code. 11487097
5000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11487097

Brand: Thermo Scientific Alfa Aesar A14445.0I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-91-5
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002516
InChI Key QQVIHTHCMHWDBS-UHFFFAOYSA-N
Synonym isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fjShow MoreShow Less
PubChem CID 8496
ChEBI CHEBI:30802
IUPAC Name benzene-1,3-dicarboxylic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 344°C to 348°C (sublimation)
Density 1.54
Quantity 5000 g
Beilstein 1909332
Merck Index 14,5197
Solubility Information Insoluble in water.
Formula Weight 166.13
Percent Purity 99%
Chemical Name or Material Isophthalic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY11K009 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberA14445.0B,A14445.30,A14445.0I
Lot NumberY15K048 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberA14445.0B,A14445.30,A14445.0I
Lot NumberU14K009 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberA14445.0B,A14445.30,A14445.0I
Lot NumberY30J001 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog NumberA14445.0B,A14445.30,A14445.0I
Lot NumberU24I003 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog NumberA14445.0B,A14445.30,A14445.0I

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isophthalic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.