missing translation for 'onlineSavingsMsg'
Learn More

Isophytol, 95%, Thermo Scientific Chemicals

Product Code. 11408517
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-32-8
Molecular Formula C20H40O
Molecular Weight (g/mol) 296.539
MDL Number MFCD00048380
InChI Key KEVYVLWNCKMXJX-UHFFFAOYSA-N
Synonym isophytol, 3,7,11,15-tetramethyl-1-hexadecen-3-ol, 1-hexadecen-3-ol, 3,7,11,15-tetramethyl, 2,6,10,14-tetramethylhexadec-15-en-14-ol, 2,6,10-trimethyl-14-vinylpentadecan-14-ol, 1-hexadecene-3-ol, 3,7,11,15-tetramethyl, 1-phyten-3-ol, iso-phytol, dsstox_cid_5474
PubChem CID 10453
IUPAC Name 3,7,11,15-tetramethylhexadec-1-en-3-ol
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)(C=C)O

Specifications

Density 0.8519
Boiling Point 125°C to 128°C (0.06 mmHg)
Refractive Index 1.4571
Quantity 10 g
Beilstein 1783243
Merck Index 14,5198
Formula Weight 296.54
Percent Purity 95%
Chemical Name or Material Isophytol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'