missing translation for 'onlineSavingsMsg'
Learn More

Isoprenaline hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11683009
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-30-9
Molecular Formula C11H18ClNO3
Molecular Weight (g/mol) 247.72
MDL Number MFCD00012603,MFCD00064548
InChI Key IROWCYIEJAOFOW-UHFFFAOYNA-N
Synonym isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
PubChem CID 5807
IUPAC Name 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 165.0°C to 175.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble in cold water.
Formula Weight 247.72
Percent Purity 98%
Chemical Name or Material Isoprenaline hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'