Learn More
Isopropanol, 99.5+%, pure, Thermo Scientific Chemicals
50.25 € - 545.72 €
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
MDL Number | MFCD00011674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10215390
|
Thermo Scientific Acros
149320010 |
1 L | Glass Bottle |
50.25 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173240
|
Thermo Scientific Acros
149320025 |
2.5 L | Glass Bottle |
88.34 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10399752
|
Thermo Scientific Acros
149320050 |
5 L | Plastic Drum |
149.75 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10592921
|
Thermo Scientific Acros
149320250 |
25 L | Plastic Drum |
545.72 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Specifications
-89.5°C | |
0.7850g/mL | |
12°C (53.6°F) | |
Liquid | |
99.5+% | |
0.785 | |
(CH3)2CHOH | |
Vapor Pressure: 43mmHg at 20°C | |
01, 360 | |
13,108 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1 | |
Pure |
67-63-0 | |
81.0°C to 83.0°C | |
Authentic | |
1.3760 to 1.3780 | |
0.005 % max. | |
C3H8O | |
MFCD00011674 | |
2.27 mPa.s (20°C) | |
0.2% Max. (K.F.) | |
15, 5254 | |
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99.5% | |
Isopropanol, 99.8+% |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Isopropanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only