missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropanol, Extra Pure, SLR, Fisher Chemical™
33.30 € - 458.00 €
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
MDL Number | 11674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11428782
|
Fisher Chemical
P/7490/08 |
500 mL | Amber Glass Bottle |
33.30 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12698645
|
Fisher Chemical
P/7490/PB08 |
500 mL | Plastic-coated Glass Bottle |
42.70 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10477070
|
Fisher Chemical
P/7490/15 |
1 L | Amber Glass Bottle |
82.80 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10588630
|
Fisher Chemical
P/7490/17 |
2.5 L | Amber Glass Bottle |
176.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10284200
|
Fisher Chemical
P/7490/PB17 |
2.5 L | Plastic-coated Glass Bottle |
194.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10070320
|
Fisher Chemical
P/7490/21 |
5 L | Plastic Bottle |
213.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10595729
|
Fisher Chemical
P/7490/FP21 |
5 L | Plastic Container |
146.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306390
|
Fisher Chemical
P/7490/25 |
25 L | Metal Can |
458.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10731974
|
Fisher Chemical
P/7490/PC25 |
25 L | Plastic Container |
389.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Isopropanol, Extra Pure, SLR
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
11674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Specifications
7 | |
67-63-0 | |
81°C | |
C3H8O | |
43mmHg at 20°C | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 |
-89.5°C | |
Colorless | |
Liquid | |
11674 | |
2.27 mPaS at 20°C | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Propan-2-ol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.