missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Optima™LC/MS Grade, Fisher Chemical™

Product Code. 10684355
2.5 L, Glass Bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number 11674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Absorbance Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm
Color Colorless
Density 0.785g/cm³
Boiling Point 81°C
Quantity 2.5 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range Assay: ≥99.90%
Residue after Evaporation 1ppm max.
Linear Formula (CH3)2CHOH
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPaS at 20°C
Water 0.05% max.
Merck Index 15, 5254
Formula Weight 60.1
Percent Purity ≥99.90%
Grade Also meets ACS specifications
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iso-propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Iso-propanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.