missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl acetate, 99%, pure, Thermo Scientific Chemicals

Product Code. 10604632
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-21-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00008877
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Synonym isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C

Specifications

Melting Point -73.0°C
Color Colorless
Density 0.8720g/mL
Boiling Point 88.8°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH(CH3)2
Refractive Index 1.3760 to 1.3780
Quantity 2.5 L
Beilstein 02, 130
Merck Index 15, 5251
Specific Gravity 0.872
Solubility Information Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents
Viscosity 0.49 mPa.s (25°C)
Formula Weight 102.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Isopropyl acetate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isopropyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'