Learn More
Isopropyl-β-D-thiogalactopyranoside (IPTG) (Dioxane-free), Fisher BioReagents™
16.54 € - 284.53 €
Chemical Identifiers
CAS | 367-93-1 |
---|---|
Molecular Formula | C9H18O5S |
Molecular Weight (g/mol) | 238.30 |
MDL Number | MFCD00063273 |
InChI Key | BPHPUYQFMNQIOC-NXRLNHOXSA-N |
Synonym | iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactosideShow More |
PubChem CID | 656894 |
ChEBI | CHEBI:61448 |
SMILES | CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10214633
|
Fisher BioReagents
BP1755-1 |
1 g | Glass Bottle | N/A | N/A | N/A | |||||
10356553
|
Fisher BioReagents
BP1755-10 |
10 g | Glass Bottle | N/A | N/A | N/A | |||||
10021793
|
Fisher BioReagents
BP1755-100 |
100 g | Poly Bottle | N/A | N/A | N/A | |||||
Description
Chemical Identifiers
367-93-1 | |
238.30 | |
BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
656894 | |
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
C9H18O5S | |
MFCD00063273 | |
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactosideShow More | |
CHEBI:61448 |
Specifications
367-93-1 | |
White | |
≥99 % (by TLC) | |
MFCD00063273 | |
-28.5 to -34.5° (+ or −) | |
BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol | |
656894 | |
238.31 | |
Solid |
110° to 115°C | |
5 to 7 | |
C9H18O5S | |
15, 5126 | |
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
238.30 | |
CHEBI:61448 | |
≥99% | |
Isopropyl-β-D-thiogalactopyranoside, Dioxane-free |
Safety and Handling

CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:1
Recommended Storage : 4°C
- Isopropyl-ß-D-thiogalactopyranoside
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.