Learn More
Isopropyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Acidity | 0.005% max. |
Density | 0.7200g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Glass bottle |
Molecular Formula | C6H14O |
Linear Formula | [(CH3)2CH]2O |
Refractive Index | 1.3670 to 1.3690 (20°C, 589nm) |
Quantity | 1 L |
UN Number | 1159 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466995 | Certificate of Analysis | 01/11/2024 | 180680025,180680010,180680250 |
A0459715 | Certificate of Analysis | 02/02/2024 | 180680025,180680010,180680250 |
B0542398 | Certificate of Analysis | 14/11/2023 | 180680025,180680010,180680250 |
B0539347A | Certificate of Analysis | 20/10/2023 | 180680025,180680010,180680250 |
B0540025A | Certificate of Analysis | 20/10/2023 | 180680025,180680010,180680250 |
Safety and Handling


- Isopropyl ether
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.