missing translation for 'onlineSavingsMsg'
Learn More
Isopropyl formate, 98%, Thermo Scientific Chemicals Product Code.: 11463447

Isopropyl formate, 98%, Thermo Scientific Chemicals

Product Code. 11463447
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11463447

Brand: Thermo Scientific Alfa Aesar A17222.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isopropyl formate, is a flavoring agent. It can also be used as fragrance agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl formate, is a flavoring agent. It can also be used as fragrance agent.

Solubility
slightly soluble in water, completely miscible with alcohol, ether, and most organic solvents.

Notes
d^2^0 0.901. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-55-8
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00014127
InChI Key RMOUBSOVHSONPZ-UHFFFAOYSA-N
Synonym isopropyl formate, formic acid, 1-methylethyl ester, formic acid isopropyl ester, formic acid, isopropyl ester, isopropyl methanoate, 1-methylethyl formate, methylethyl formate, unii-h1l164w42g, fema no. 2944, hcooch ch3 2
PubChem CID 12257
IUPAC Name propan-2-yl formate
SMILES CC(C)OC=O

Specifications

Melting Point -93°C
Density 0.88
Boiling Point 68°C to 71°C
Flash Point −6°C (21°F)
Linear Formula HCO2CH(CH3)2
Refractive Index 1.3685
Quantity 250 g
UN Number UN1281
Beilstein 1735844
Solubility Information slightly soluble in water,completely miscible with alcohol,ether,and most organic solvents.
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material Isopropyl formate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241095 Certificate TypeCertificate of Analysis Date09/06/2022 Catalog NumberA17222.18,A17222.30
Lot Number10158826 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17222.18,A17222.30
Lot Number10172874 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17222.18,A17222.30
Lot NumberH4513A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17222.18,A17222.30
Lot Number10206541 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17222.18,A17222.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isopropyl formate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.