missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl salicylate, 99%, Thermo Scientific Chemicals

Product Code. 10001811
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Isopropyl salicylate, 99%, ACROS Organics, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 122.0 deg.C (18.0 mmHg), Colorless to Yellow, Molecular Weight: 180.203, Percent Purity: 99%, Assay Percent Range: 99%, CAS: 607-85-2, ChEBI: CHEBI:38703, Density: 1.0600g/mL, EINECS Number: 210-143-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 607-85-2
MDL Number MFCD00035703
InChI Key YEULQIJMIOWCHB-UHFFFAOYSA-N
Synonym isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate
PubChem CID 11838
ChEBI CHEBI:38703
IUPAC Name propan-2-yl 2-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=CC=C1O

Spezifikation

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 122.0°C (18.0 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C10H12O3
Refractive Index 1.5090 to 1.5110
Quantity 100 g
Specific Gravity 1.06
Molecular Weight (g/mol) 180.2
Formula Weight 180.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isopropyl salicylate, 99%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Isopropyl salicylate
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'