Learn More
Isopropylamine, 99%, Thermo Scientific Chemicals
24.60 € - 43.70 €
Chemical Identifiers
CAS | 75-31-0 |
---|---|
Molecular Formula | C3H9N |
Molecular Weight (g/mol) | 59.11 |
InChI Key | JJWLVOIRVHMVIS-UHFFFAOYSA-N |
Synonym | isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino |
PubChem CID | 6363 |
ChEBI | CHEBI:15739 |
IUPAC Name | propan-2-amine |
SMILES | CC(C)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10153050
|
Thermo Scientific Acros
148920250 |
25 mL | Glass bottle |
24.60 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10667112
|
Thermo Scientific Acros
148920010 |
1 L | Glass bottle |
43.70 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-31-0 | |
59.11 | |
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino | |
CHEBI:15739 | |
CC(C)N |
C3H9N | |
JJWLVOIRVHMVIS-UHFFFAOYSA-N | |
6363 | |
propan-2-amine |
Specifications
75-31-0 | |
White | |
33.0°C to 34.0°C | |
Authentic | |
C3H9N | |
1.3725 to 1.3755 | |
13,164 | |
0.69 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol and ether | |
CC(C)N | |
59.11 | |
CHEBI:15739 | |
59.11 | |
Crystalline Powder, Crystals or Needles |
-101.0°C | |
0.6900g/mL | |
−37°C | |
98.5% min. (GC) | |
(CH3)2CHNH2 | |
04,152 | |
15,5255 | |
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino | |
JJWLVOIRVHMVIS-UHFFFAOYSA-N | |
propan-2-amine | |
6363 | |
0.3 mPa.s (20°C) | |
99% | |
Isopropylamine |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-860-9
- Isopropylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
- H224-Extremely flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only