missing translation for 'onlineSavingsMsg'
Learn More

Isoproterenol hydrochloride, Tocris Bioscience™

Product Code. 30000721
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Standard selective β agonist

Standard selective β-adrenoceptor agonist; vasorelaxant and bronchodilator. Activation of β2 receptors activates downstream PKA and ERK, and may stimulate NO-mediated endothelium-dependent smooth muscle relaxation. Active in vivo.

Specifications

CAS 51-30-9
Molecular Formula C11H18ClNO3
MDL Number MFCD00012603,MFCD00064548
Quantity 100 mg
Synonym isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
InChI Key IROWCYIEJAOFOW-UHFFFAOYNA-N
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1
IUPAC Name hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride
Molecular Weight (g/mol) 247.72
PubChem CID 5807
Formula Weight 247.72
Percent Purity >97%
Chemical Name or Material Isoproterenol hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.