missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Isorack Starter Set
Ideal for cooling, transporting, and storing deep-frozen samples. Isorack is part of the Eppendorf IsoTherm System
Brand: Eppendorf™ 5825721004
This item is not returnable.
View return policy
Description
- IsoTherm-System: IsoRack (work rack), IsoSafe (insulating box), and two IsoPacks (cool packs)
- Dry incubation technology reduces contamination risk associated with classical icebath incubation for safe sample handling
- Holds up to 24 micro tubes (e.g., 0.5 mL or 1.5 mL/2.0 mL) for broad flexibility
- Two versions of IsoPack (cold pack) available: White maintains 0 °C for up to 6 hours. Blue maintains -21 °C for up to 3 hour
- Racks are stackable, autoclavable and can be centrifuged in the MTP rotor
Specifications
Isorack Starter Set | |
Flex Bucket |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction