missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide dimethyl ether, 99%, Thermo Scientific Chemicals

Catalog Number 10764753
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5306-85-4
C8H14O4
174.20
MFCD00064828
MEJYDZQQVZJMPP-UHFFFAOYNA-N
dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan
62990
COC1COC2C(COC12)OC
This item is not returnable. View return policy
5306-85-4
C8H14O4
174.20
MFCD00064828
MEJYDZQQVZJMPP-UHFFFAOYNA-N
dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan
62990
COC1COC2C(COC12)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 93°C to 95°C (0.1 hPa)
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4600 to 1.4620
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.17
Beilstein 19, IV, 991
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
IUPAC Name 3,6-dimethoxy-hexahydrofuro[3,2-b]furan
Formula Weight 174.19
Percent Purity 99%
Chemical Name or Material Isosorbide dimethyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'