Learn More
Isosorbide mononitrate, MedChemExpress
MedChemExpress Isosorbide mononitrate(Isosorbide-5-mononitrate) is a nitrate-class compound used for angina pectoris; acts by dilating the blood vessels so as to reduce the blood pressure.

110.70 € - 248.40 €
Specifications
Chemical Name or Material | Isosorbide mononitrate |
---|---|
CAS | 16051-77-7 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C6H9NO6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16492249
![]() |
Medchem Express
HY-B0642 |
1 g |
110.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16491028
![]() |
Medchem Express
HY-B0642 |
5 g |
248.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isosorbide mononitrate(Isosorbide-5-mononitrate) is a nitrate-class compound used for angina pectorisSpecifications
Isosorbide mononitrate | |
White | |
C6H9NO6 | |
DMSO : 100 mg/mL (523.18 mM; Need ultrasonic) ∣H2O : 50 mg/mL (261.59 mM; Need ultrasonic) | |
191.14 | |
99.57% | |
Solid |
16051-77-7 | |
Research | |
Isosorbide-5-mononitrate | |
O=[N+]([O-])O[C@H]1[C@](OC[C@@H]2O)([H])[C@]2([H])OC1 | |
191.14 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H228∣H361
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.