missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp II™ Recirculating Chiller, 230V/50Hz

A compact line of refrigerated recirculating chillers.

Brand:  Fisherbrand™ 197222010001

3360.69 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 11326641

  • 4185.00 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Intuitive digital controller for ease of use
  • Pressure gauge on front panel for easy viewing of application pressure
  • High and low temperature alarms
Controller indicator lights:
  • Indicate when unit is cooling
  • Indicate when setpoint is being changed
  • Indicate when high and low temperature limits are being changed or are exceeded
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10°C to 35°C
900 W (at +20°C)
-10°C to +30°C
5.3 L/min. (at 4.1 bar)
±0.1°C
230 V
9.5 L
PD1 (Positive Displacement Pump)
Isotemp II
R134A
584.2 x 355.6 x 604.5 mm
50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers