missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ IsoTherm-System™

Product Code. 10613751 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
0.5mL tubes
1.5/2.0mL tubes
Type:
IsoPack and IsoRack set
IsoSafe and IsoPack
Temperature Range:
-21°C
0°C
Unit Size:
Each
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 10613751

Brand: Eppendorf™ 3880000020

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Used as an enzyme bar to prevent enzymes from becoming inactivated, as well as for PCR* sample preparation, sample transport and slow thawing of frozen samples.

  • Holds 24 tubes (0.5mL/1.5mL/2.0mL Safe-Lock microcentrifuge tubes or 1.7mL Safe-Twist screw cap tubes)
  • IsoPack™ (cold pack) white for 0°C and blue for -21°C, cooled to refrigeration temperature in freezer compartment
  • Isothermal times of >6hr (0°C, white) and >3hr (-21°C, blue)
  • No more melting ice baths (danger of contamination)
  • All system components can be used separately
  • Racks are stackable, autoclavable, markable, floatable and can be centrifuged in a microtitre-plate rotor Specialises in cooling samples effectively and constantly at -21°C or 0°C over many hours.
TRUSTED_SUSTAINABILITY

Specifications

Type IsoSafe and IsoPack
Temperature Range 0°C
For Use With (Equipment) 0.5mL tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.