missing translation for 'onlineSavingsMsg'
Learn More

Isovaleryl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11406906
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-12-3
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00000738
InChI Key ISULZYQDGYXDFW-UHFFFAOYSA-N
Synonym isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
PubChem CID 66054
IUPAC Name 3-methylbutanoyl chloride
SMILES CC(C)CC(=O)Cl

Specifications

Density 0.989
Boiling Point 115°C to 117°C
Flash Point 27°C (80°F)
Refractive Index 1.416
Quantity 500 g
UN Number UN2920
Beilstein 741910
Sensitivity Moisture sensitive
Merck Index 14,5232
Formula Weight 120.58
Percent Purity 98%
Chemical Name or Material Isovaleryl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'