missing translation for 'onlineSavingsMsg'
Learn More

Isovaleryl chloride, 98%, Thermo Scientific Chemicals

25.53 € - 161.81 €

Chemical Identifiers

CAS 108-12-3
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000738
InChI Key ISULZYQDGYXDFW-UHFFFAOYSA-N
Synonym isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
PubChem CID 66054
IUPAC Name 3-methylbutanoyl chloride
SMILES CC(C)CC(=O)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10429120
View Documents Promotion Details
Thermo Scientific Acros
169280250
25 g Glass bottle
25.53 €
25g
Please to purchase this item. Need a web account? Register with us today!
10502581
View Documents Promotion Details
Thermo Scientific Acros
169281000
100 g Glass bottle
45.53 €
100g
Please to purchase this item. Need a web account? Register with us today!
10012070
View Documents Promotion Details
Thermo Scientific Acros
169285000
500 g Glass bottle
161.81 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-12-3
120.58
ISULZYQDGYXDFW-UHFFFAOYSA-N
66054
CC(C)CC(=O)Cl
C5H9ClO
MFCD00000738
isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
3-methylbutanoyl chloride

Specifications

108-12-3
0.9800g/mL
27°C
98%
(CH3)2CHCH2COCl
MFCD00000738
15, 5278
isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
ISULZYQDGYXDFW-UHFFFAOYSA-N
3-methylbutanoyl chloride
66054
98%
Isovaleryl chloride, 98%
Colorless to Yellow or Pink
115°C to 117°C
Authentic
C5H9ClO
1.4150 to 1.417
02, 315
0.98
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol
CC(C)CC(=O)Cl
120.58
120.58
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-552-2

missing translation for 'tsca' : TSCA

Product Identifier
  • Isovaleryl chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only