missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carbothioamide, 97%, Thermo Scientific™

Product Code. 10641503
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175334-72-2
Molecular Formula C4H4N2OS
Molecular Weight (g/mol) 128.15
MDL Number MFCD00276995
InChI Key CEDRGDFENMZKCQ-UHFFFAOYSA-N
Synonym isoxazole-5-carbothioamide, 5-isoxazolecarbothioamide, isoxazole-5-carbothioic acid amide, 5-isoxazolecarbothioamide 9ci, aminoisoxazol-5-ylmethane-1-thione
PubChem CID 2799515
IUPAC Name 1,2-oxazole-5-carbothioamide
SMILES NC(=S)C1=CC=NO1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 128.15
Percent Purity 97%
Chemical Name or Material Isoxazole-5-carbothioamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.