missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoxazole-5-carboxylic acid, 95%, Thermo Scientific™
20.90 € - 59.60 €
Chemical Identifiers
CAS | 21169-71-1 |
---|---|
Molecular Formula | C4H3NO3 |
Molecular Weight (g/mol) | 113.072 |
MDL Number | MFCD00156151 |
InChI Key | MIIQJAUWHSUTIT-UHFFFAOYSA-N |
Synonym | isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h |
PubChem CID | 2060599 |
IUPAC Name | 1,2-oxazole-5-carboxylic acid |
SMILES | C1=C(ON=C1)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10254964
|
Thermo Scientific Maybridge
CC10301DA |
1 g |
20.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10732357
|
Thermo Scientific Maybridge
CC10301DE |
5 g |
59.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
21169-71-1 | |
113.072 | |
MIIQJAUWHSUTIT-UHFFFAOYSA-N | |
2060599 | |
C1=C(ON=C1)C(=O)O |
C4H3NO3 | |
MFCD00156151 | |
isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h | |
1,2-oxazole-5-carboxylic acid |
Specifications
21169-71-1 | |
100.0 | |
Amber glass bottle | |
MFCD00156151 | |
MIIQJAUWHSUTIT-UHFFFAOYSA-N | |
1,2-oxazole-5-carboxylic acid | |
2060599 | |
97% |
96.5 | |
97% | |
C4H3NO3 | |
isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h | |
C1=C(ON=C1)C(=O)O | |
113.072 | |
113.07 | |
Isoxazole-5-carboxylic acid |