missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11464764
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reaction conditions were found to allow the exclusive formation of isoxazole-5-carboxylic acid derivatives by conjugate addition, in acidic medium, of hydroxylamine to β-alkoxyvinyl trichloromethyl ketone in synthesis and reactivity of isoxazoles. Isoxazole-5-carboxylic acid participates in the Synthesis and evaluation of acylguanidine FXa inhibitors. 1, 1, 1-trichloro-4-methoxy-3-penten-2-one postulated in the preparation of isoxazole-5-carboxylic acid from trichloroacetyl chloride, ethyl vinyl ether and hydroxylamine is developed in a fully saturated nitrogen atom theerefore there is no possibility of conjugation with 4- substituents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reaction conditions were found to allow the exclusive formation of isoxazole-5-carboxylic acid derivatives by conjugate addition, in acidic medium, of hydroxylamine to β-alkoxyvinyl trichloromethyl ketone in synthesis and reactivity of isoxazoles. Isoxazole-5-carboxylic acid participates in the Synthesis and evaluation of acylguanidine FXa inhibitors. 1, 1, 1-trichloro-4-methoxy-3-penten-2-one postulated in the preparation of isoxazole-5-carboxylic acid from trichloroacetyl chloride, ethyl vinyl ether and hydroxylamine is developed in a fully saturated nitrogen atom theerefore there is no possibility of conjugation with 4- substituents.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21169-71-1
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.072
MDL Number MFCD00156151
InChI Key MIIQJAUWHSUTIT-UHFFFAOYSA-N
Synonym isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h
PubChem CID 2060599
IUPAC Name 1,2-oxazole-5-carboxylic acid
SMILES C1=C(ON=C1)C(=O)O

Specifications

Melting Point 141°C to 144°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 113.07
Percent Purity 98%
Chemical Name or Material Isoxazole-5-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'