missing translation for 'onlineSavingsMsg'
Learn More

Itaconic acid, 99+%

Product Code. 11945031 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11945031

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-65-4 | C5H6O4 | 130.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-65-4
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.1
InChI Key LVHBHZANLOWSRM-UHFFFAOYSA-N
Synonym itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid
PubChem CID 811
ChEBI CHEBI:30838
IUPAC Name 2-methylidenebutanedioic acid
SMILES C=C(CC(=O)O)C(=O)O

Specifications

Melting Point 165.0°C to 168.0°C
Color White to Beige
Density 1.5700g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic Bottle
Linear Formula HO2CCH2C(=CH2)CO2H
Quantity 5 kg
Beilstein 02,76
Merck Index 15,529
Specific Gravity 1.57
Solubility Information Solubility in water: 95g/L (25°C). Other solubilities: soluble in alcohol and acetone,slightly soluble in most other common organic,solvents
Formula Weight 130.1
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Itaconic acid
Show More Show Less
compliance-icons
Product Identifier
  • Itaconic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Itaconic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.