missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Itraconazole, 99%

Product Code. 15279786 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15279786

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Itraconazole, or voriconazole, CAS # 84625-61-6, is an N-arylpiperazine that has antifungal properties. | CAS: 84625-61-6 | C35H38Cl2N8O4 | 705.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Itraconazole is an aromatic ether that works as a strong inhibitor of P-glycoproteins and CYP3A4

• Itraconazole also acts as a hedgehog signaling inhibitor and P450 inhibitor

Applications

• Itraconazole can be exhibits antifungal activity against organisms including aspergillosis, blastomycosis, candidiasis, chromoblastomycosis, coccidioidomycosis, cryptococcosis, histoplasmosis, and sporotrichosis

• Itraconazole may be used to experimentally target the delivery of other molecules, such as plasmids, to cells

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84625-61-6
Molecular Formula C35H38Cl2N8O4
Molecular Weight (g/mol) 705.64
MDL Number MFCD00870168,MFCD00941396
InChI Key VHVPQPYKVGDNFY-ZPGVKDDISA-N
Synonym itraconazole-d5, itraconazole, oriconazole-d5, triasporin-d5, sporamelt-d5, itrizole-d5, sporanox-d5, +/--4-4-4-4-2r,4s-2,4-dichlorophenyl-2-1h-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl-1-piperazinyl phenyl-2,4-dihydro-2-1-methylpropyl-d5-3h-1,2,4-triazol-3-one, 2h5-4-4-4-4-2-1h-1,2,4-triazol-1-yl methyl-2-2,4-dichlorophenyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1-sec-butyl-1h-1,2,4-triazol-5 4h-one, 2-3,3,4,4,4-2 h? butan-2-yl-4-4-4-4-2r,4s-2-2,4-dichlorophenyl-2-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1,2,4-triazol-3-one
PubChem CID 45039617
IUPAC Name 4-[4-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-(3,3,4,4,4-pentadeuteriobutan-2-yl)-1,2,4-triazol-3-one
SMILES CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1

Specifications

Melting Point 168.0°C to 170.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 5 g
Merck Index 15,5292
Solubility Information Solubility in water: <1mg/mL. Other solubilities: solubility in ethanol: <1mg/mL, solubility in dmso: 4mg/mL
Formula Weight 705.64
Percent Purity 99%
Chemical Name or Material Itraconazole
compliance-icons
Product Identifier
  • Itraconazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.