missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Standard IUPAC pH 1.679 at 25°C
Click to view available options
Quantity:
500 mL
Description
- IUPAC pH Standard, pH 1.679
- Radiometer pH buffer solution supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
- Total Control of the traceability chain and calculated uncertainties
- Direct traceability to primary standards and the reference method
- The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
Specifications
Specifications
pH | 1.679 |
Physical Form | Liquid |
Quantity | 500 mL |
Traceability to NIST | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'