missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IUPAC pH Standard pH 9.180, Hach Lange™
Click to view available options
Quantity:
500 mL
Description
- pH Standard, IUPAC, pH9.180 at 25 °C, Certified, 500 mL (Radiometer Analytical)
- Each bottle is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31 500 mL
- Features a concentration of 9.180 ± 0.010 pH at 25°C
- Traceable to IUPAC pH scale and NIST
- Shelf life of 48 months from production date
- The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
Specifications
Specifications
pH | 9.180 |
Physical Form | Liquid |
Quantity | 500 mL |
Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction