missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulphuric acid 95-97%, BAKER ANALYZED™, J.T. Baker™
Laboratory Reagent
57.00 € - 140.00 €
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10668151
|
JT Baker
6057.1000 |
1 L | Glass bottle |
57.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10506812
|
JT Baker
6057.2500 |
2.5 L | Glass bottle |
140.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10036110
|
JT Baker
6057.2500PE |
2.5 L | Plastic bottle |
110.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
OS(O)(=O)=O |
H2O4S | |
MFCD00064589 | |
sulfuric acid |
Specifications
7664-93-9 | |
1.84 g/cm³ (20°C) | |
max. 5 ppm | |
MFCD00064589 | |
OS(O)(=O)=O | |
98.07 |
3°C | |
max. 5 ppm | |
H2O4S | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- sulphuric acid
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title