Learn More
Avantor™ J.T. Baker™ BAKERBOND spe™ SDB-1 SPE columns, polymer based
Can improve the quality of sample preparation
Brand:  Avantor™ 7519-02
Product Code. 10331281
  • 249.00 € / Pack of 50
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent structural, mechanical and chemical properties, packaging purity and load ability, as well as stability of the SDB sorbent, offer improved sample clean-up and pre-concentration.
  • This SDB polymeric sorbent is a highly efficient and selective SDB sorbent with high throughput performance.
  • It enables high recovery and reproducibility of results, even in the SPE of compounds of different polarities.
Specifications
Specifications
3 mL
200 mg
50