Learn More
Avantor™ J.T. Baker™ BAKERBOND Speedisk™ Expanded Extraction Station
Can be used in reservoir, inverted, or remote sample feed modes
Brand:  Avantor™ 8095-06
Product Code. 15597694
  • 7125.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • It includes a six-port vacuum manifold and the accessories needed to support the extraction of analyte by BAKERBOND Speedisk™ laminar extraction discs.
  • The manifold has a rectangular footprint and inter-port spacing to accommodate six, side-by-side, 1 L sample reservoirs
  • Each vacuum port has an individual open/close valve.
  • Includes extraction station, two remote sample adapters, two collection chambers and two vials, six reservoirs of 185 mL
Specifications
Specifications
6
185 mL
Expanded