Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum is a gas collecting tube with O-ring valves on each end and a centrally located septum.
172.00 € - 219.00 €
Specifications
Type Gas Sample Tube
View More Specs
Products 6
Product Code Brand Measurement Range Price Quantity & Availability  
Product Code Brand Measurement Range Price Quantity & Availability  
15874764
View Documents
GPE Scientific™
GSS/100
100 mL
172.00 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
15894764
View Documents
GPE Scientific™
GSS/125
125 mL
173.00 €
Each
Estimated Shipment: 31-12-2024
to see stock.
Add to basket
15824774
View Documents
GPE Scientific™
GSS/500
500 mL
186.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
15814774
View Documents
GPE Scientific™
GSS/250
250 mL
181.00 €
Each
Estimated Shipment: 20-12-2024
to see stock.
Add to basket
15884764
View Documents
GPE Scientific™
GSS/1000
1000 mL
195.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
15804774
View Documents
GPE Scientific™
GSS/2000
2000 mL
219.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
Description
Description
  • Cylindrical, oblong shape
  • Gas tight container
  • Gauged volume
  • Standard collecting tube with valved ends
  • Includes centralized septum
Specifications
Specifications
Gas Sample Tube