Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap
Product Code. 15814794
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 15814794
Brand: GPE Scientific™ JCF/1000
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap is specially designed for use with vacuum manifolds and Schlenk lines.
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap
Specifications
Capacity (Metric) 1000 mL
For Use With (Equipment) Vacuum manifolds and Schlenk lines
Includes Fine thread tap
Product Type Sample Flask